Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK beta 2/PRKAB2 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4H4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271226AF647

Catalog No. N271226A647


Add to cart

Description

Description

AMPK beta 2/PRKAB2 Monoclonal antibody specifically detects AMPK beta 2/PRKAB2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

AMPK beta 2/PRKAB2
OTI4H4
Western Blot
Affinity Purified, Conjugated
PRKAB2
Full length recombinant protein of human AMPK beta 2/PRKAB2 (NP_005390) produced in E.coli.
Immunogen affinity purified
Lipid and Metabolism
Primary
5565.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
5′-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5′-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, protein kinase, AMP-activated, beta 2 non-catalytic subunit
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit