Learn More
MBL International Corporation CycLex AMPK Kinase Assay Kit
SDP
Supplier:  MBL International Corporation CY1182
Encompass_Preferred
Size:96 Assays; Target:AMPK; The AMPK kinase assay kit can be used to measure the activity of AMPK for 96 assays. Applications of the kit include screening activators or inhibitors of AMPK, evaluating the effects of pharmacological agents on AMPK in vitro or monitoring the effects of purification of AMPK. MBL International's AMPK kinase assay kit is highly cited.Bulk pricing available.CycLex AMPK Kinase Assay Kit.
Catalog No. 50-455-68
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.