Learn More
Aat BioquestSupplier Diversity Partner Amplite™ Red  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11011
Our Amplite™ Red is a sensitive fluorogenic peroxidase substrate that generates a highly red fluorescent product that has maximum absorption of 571 nm and maximum emission of 585 nm.
Catalog No. 50-195-3333