Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amprolium, USP, 97-101%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle
121-25-5
C14H19ClN4
278.78
LCTXBFGHZLGBNU-UHFFFAOYSA-M
1-[(4-amino-2-propylpyrimidin-5-yl)methyl]-2-methylpyridin-1-ium chloride
[Cl-].CCCC1=NC=C(C[N+]2=CC=CC=C2C)C(N)=N1
121-25-5
C14H19ClN4
278.78
LCTXBFGHZLGBNU-UHFFFAOYSA-M
1-[(4-amino-2-propylpyrimidin-5-yl)methyl]-2-methylpyridin-1-ium chloride
[Cl-].CCCC1=NC=C(C[N+]2=CC=CC=C2C)C(N)=N1

A2728, 121-25-5, C14H19ClN4

Spectrum™ Amprolium, USP is used as a coccidiostat in poultry. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.01
Quantity 1 kg
CAS Max % 1
Percent Purity 97 to 101%