Thermo Scientific™ AMS-4 Beta Air Monitor Available on GSA/VA Contract for Federal Government customers only.

Provide an early warning to workers exposed to potential airborne releases of beta-emitting particulates when using Thermo Scientific™ AMS-4 Beta Air Monitor.

Manufacturer: Thermo Scientific™ AMS4OPT13

 View more versions of this product

Catalog No. 19-175-231

  Check Availability
Add to Cart

    Specifications

    Power Consumption 1.5A
    Description System Monitoring and Calibration Software with Cables
    Power 0.2hp
    Certifications/Compliance Meets Regulatory Guide 8.25 and revised 10 CFR 20 criteria
    Compatibility Thermo Scientific ViewPoint Enterprise
    Item Description Monitoring and calibration software
    Voltage 220V

    Provide an early warning to workers exposed to potential airborne releases of beta-emitting particulates when using Thermo Scientific™ AMS-4 Beta Air Monitor. Lightweight and robust design accommodates both fixed and portable use applications.

    Continuous air monitor (CAM) offering beta particulate, radioiodine, or noble gas detection.

    • Detachable sampling head
    • Reports both DAC and cpm
    • Real time gamma background subtraction
    • Stand alone or network configuration
    • Mass flow measurements
    • Fast and slow concentration alarms, cpm alarm, DAC-hour alarm
    • Computer calibration
    • Can also be used for stack monitoring of effluent releases using the in-line detector head
    • Keypad: 18 keys, full numeric plus function keys
    • Status Indicators: Front panel lights display READY and MALFUNCTION conditions, red alarm strobe light, sonalert
    • Printer: RS-232 serial DB9
    • Data Log Buffer: 2000 entries of combined Concentration and Status
    • Change information: Equivalent to 83 days of 1 hour detail, nearly 14 days of 10 mintes detail or over 24 hours of 1 minute detail
    • Alarm/Fail Relays: SPST, 15A at 30VDC/120VAC resistive
    • Analog Output: 0 to 5VDC output, five decade logarithmic

    Compatible with:

    Thermo Scientific ViewPoint Enterprise

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit