Learn More
BioworldSupplier Diversity Partner AMV (Avian Myeloblastosis Virus) Reverse Transcriptase, 200 Unit(s) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600071
- Source: Avian Myeloblastosis Virus(AMV) AMV Rev Trans Activity(U/uL)- 5-10 AMV Rev Trans Nuclease(P/F)- Pass First Strand cDNA Synthesis(P/F)- Pass
Catalog No. 50-253-099