Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate, 98%, Spectrum™

Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
628-63-7
628-63-7
Packaging Amber Glass Bottle
Acidity 0.0001
Water 0.002
Quantity 100 mL
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.