Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate (Laboratory), Fisher Chemical™

Catalog Number A718500
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
628-63-7
C7H14O2
130.187
MFCD00009500
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C
628-63-7
C7H14O2
130.187
MFCD00009500
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C
Melting Point -70.8°C
Color Colorless
Density 0.870g/cm³
Boiling Point 149°C
Flash Point 40°C
Packaging Glass Bottle
Linear Formula CH3(CH2)4OC(O)CH3
Quantity 500 mL
UN Number 1104
Formula Weight 130.19
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Amyl Acetate
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit