Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate, Mixed Isomers, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
628-63-7
C7H14O2
130.187
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate,n-amyl acetate,acetic acid, pentyl ester,n-pentyl acetate,amyl acetic ester,1-pentyl acetate,n-pentyl ethanoate,birnenoel,pent-acetate,amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C
628-63-7
C7H14O2
130.187
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate,n-amyl acetate,acetic acid, pentyl ester,n-pentyl acetate,amyl acetic ester,1-pentyl acetate,n-pentyl ethanoate,birnenoel,pent-acetate,amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C

High quality chemicals for laboratory and specialized industrial use

Flash Point 33.00°C
Specific Gravity 1L = 0.81kg
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL
Formula Weight 88.15
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Amyl Acetate, Mixed Isomers