Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate, Natural, 95%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
628-63-7
628-63-7
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 1
Percent Purity 0.95
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.