Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Acetate, Technical, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
628-63-7
628-63-7
Packaging Steel Can
Quantity 20 L
CAS Max % 1
Grade Technical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.