Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Methyl Sulfide 97.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
1741-83-9
C6H14S
118.24
MFCD00027274
FOJGPFUFFHWGFQ-UHFFFAOYSA-N
15620
1-(methylsulfanyl)pentane
CCCCCSC
1741-83-9
C6H14S
118.24
MFCD00027274
FOJGPFUFFHWGFQ-UHFFFAOYSA-N
15620
1-(methylsulfanyl)pentane
CCCCCSC
Color Yellow
Melting Point -94°C
Boiling Point 145°C
UN Number 1993
Quantity 5mL
Formula Weight 118.24
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material Amyl Methyl Sulfide