Learn More
Thermo Scientific™ Amylase Reagent
Determine quantity of amylase in serum or urine in vitro with Thermo Scientific™ Amylase Reagent, suitable for manual use or on automated clinical chemistry analyzers.
$979.00
Specifications
CE Marker Yes
Linearity 10 to 2000U/L
Methods Compatibility Kinetic
Stability Stable Liquid
State at Room Temperature Liquid
View More Specs
Products 1
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
TR25421
Accessories View Documents
2 x 125 mL
Pack of 2 for $979.00
Add to cart
Description
Description
Determine quantity of amylase in serum or urine in vitro with Thermo Scientific™ Amylase Reagent, suitable for manual use or on automated clinical chemistry analyzers.
Amylase is most frequently measured in the diagnosis of acute pancreatitis when serum levels may be grossly elevated. Hyperamylasemia is associated with, but not limited to, other acute abdominal disorders, biliary tract disease, diabetic ketoacidosis, severe glomerular dysfunction, salivary gland disorders, ruptured ectopic pregnancy and macroamylasemia. Applications for a wide range of automated analyzers are available on request.
Specifications
Specifications
Yes
Kinetic
Liquid
10 to 2000U/L
Stable Liquid