Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amyloid β 1-42 Mouse anti-Human, Clone: mHJ7.4, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | Amyloid β 1-42 |
---|---|
Clone | mHJ7.4 |
Concentration | 1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN242125
|
MilliporeSigma
MABN242125UG |
25 μg |
Each for $177.00
|
|
|||||
MABN2421100
|
MilliporeSigma
MABN2421100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
Amyloid β 1-42 Monoclonal antibody specifically detects Amyloid β 1-42 in Human samples. It is validated for ELISA.Specifications
Amyloid β 1-42 | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG2b κ | |
Protein G purified |
mHJ7.4 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
A linear peptide corresponding to six amino acids (37-42) from the C-terminal region of human Amyloid β1-42 peptide. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title