Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid beta fibrils Rabbit anti-Human, Clone: M64, MilliporeSigma™

Rabbit Monoclonal Antibody

$145.80 - $368.50

Specifications

Antigen Amyloid beta fibrils
Clone M64
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN637I100
View Documents
MilliporeSigma
MABN637I100UG
100 μg
Each for $368.50
Add to cart
 
MABN637I25
View Documents
MilliporeSigma
MABN637I25UG
25 μg
Each for $145.80
Add to cart
 
Description

Description

Amyloid beta fibrils Monoclonal antibody specifically detects Amyloid beta fibrils in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

Amyloid beta fibrils
Immunohistochemistry (Paraffin)
Purified
RUO
Purified rabbit recombinant monoclonal antibody in PBS with 0.05% sodium azide.
Monoclonal
Protein A purified
M64
Unconjugated
Rabbit
Human
Fibrillar Abeta 1-42 peptide.
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit