Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Amyloid Precursor Protein (APP), Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

$193.10 - $968.00

Specifications

Antigen Amyloid Precursor Protein (APP)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RB9023P0
View Documents
Thermo Scientific™
RB9023P0
100μL
Each for $193.10
Add to cart
 
RB9023P1
View Documents
Thermo Scientific™
RB9023P1
500μL
Each for $541.00
Add to cart
 
RB9023P
View Documents
Thermo Scientific™
RB9023P
1mL
Each for $968.00
Add to cart
 
RB9023R7
View Documents
Thermo Scientific™
RB9023R7
7mL
Each for $246.50
Add to cart
 
RB9023R1
View Documents
Thermo Scientific™
RB9023R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Amyloid Precursor Protein (APP), Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.


Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide corresponding to N-terminal of human APP

Molecular Weight: 110kDa

Positive Control: Brain

Cellular Localization: Cytoplasmic and extracellular

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Amyloid Precursor Protein (APP)
Polyclonal
Rabbit
Neuroscience
A synthetic peptide corresponding to N-terminal of human APP
Western Blot
Unconjugated
IVD
Primary
IgG
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit