Learn More
Amylose, Spectrum™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
9005-82-7
9005-82-7
AM230, 9005-82-7
Spectrum™ Amylose , a helical polymer, is made of α-D-glucose units and is important in plant energy storage; it can be used in industrial and food based contexts as a water binder, thickener, emulsion stabilizer, and gelling agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g