Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyltrichlorosilane 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
107-72-2
C5H11Cl3Si
205.578
MFCD00000487
KWDQAHIRKOXFAV-UHFFFAOYSA-N
pentyltrichlorosilane,amyltrichlorosilane,silane, trichloropentyl,trichloroamylsilane,pentylsilicon trichloride,amyl trichlorosilane,silane, pentyltrichloro,unii-n9gp7288zp,hsdb 887,trichloro pentyl silane
7885
trichloro(pentyl)silane
CCCCC[Si](Cl)(Cl)Cl
107-72-2
C5H11Cl3Si
205.578
MFCD00000487
KWDQAHIRKOXFAV-UHFFFAOYSA-N
pentyltrichlorosilane,amyltrichlorosilane,silane, trichloropentyl,trichloroamylsilane,pentylsilicon trichloride,amyl trichlorosilane,silane, pentyltrichloro,unii-n9gp7288zp,hsdb 887,trichloro pentyl silane
7885
trichloro(pentyl)silane
CCCCC[Si](Cl)(Cl)Cl
Color Colorless
Boiling Point 172°C
Quantity 5g
UN Number 1728
Formula Weight 205.58
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material Amyltrichlorosilane

EINECSNumber : (2)-2041

RTECSNumber : VV4725000

TSCA : Yes