Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyris Oil W.I., Spectrum™

Click to view available options
Quantity:
100 mL
500 g
500 mL
2.5 kg
50 lb.
Packaging:
Amber Glass Bottle
Drum
8015-65-4
8015-65-4
Packaging Amber Glass Bottle
Quantity 2.5 kg
Refractive Index 1.495-1.525
Specific Gravity 0.950 - 0.975
Appearance Pale Yellow
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.