Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANA 12, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
TrkB receptor antagonist. Demonstrates a 2-site mode of action, preventing activation by BDNF non-competitively (IC50 values are 45.6 nM and 41.1 μM for the high and low affinity sites respectively). Displays no effect on TrkA and TrkC in an assay of neurite outgrowth. Demonstrates antidepressant properties. Also exhibits anti-seizure effects in an ischemia-induced neonatal seizure model by blocking down-regulation of KCC2.
Specifications
Quantity | 10 mg |
Synonym | ana-12, ana 12, n-2-hexahydro-2-oxo-1h-azepin-3-yl amino carbonyl phenyl-benzo b thiophene-2-carboxamide, trkb antagonist, n-2-2-oxoazepan-3-yl carbamoyl phenyl-1-benzothiophene-2-carboxamide, maybridge1_001394, ana12, ana hplc, n-2-2-oxoazepan-3-yl carbamoyl phenyl benzo b thiophene-2-carboxamide |
Formula Weight | 407.49 |
Percent Purity | >99% |
Chemical Name or Material | ANA 12 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction