Learn More
Tocris Bioscience™ Anagrelide hydrochloride
Potent PDE3 inhibitor
Manufacturer:  Tocris Bioscience™ 2432/10
 View more versions of this product
Catalog No. 24-321-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent type III phosphodiesterase (PDE3) inhibitor (IC 50 = 36 nM). Inhibits platelet aggregation and produces potent thrombocytopenic effects via inhibition of megakaryocyte maturation.
Specifications
Specifications
6,7-Dichloro-1,5-dihydroimidazo[2,1-b]quinazolin-2(3H)-one hydrochloride
>99%
Observed MW: 292.55
Soluble to 10mM in DMSO
58579-51-4
C10H7Cl2N3O.HCl
10mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at +4°C