Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anagrelide Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
68475-42-3
C10H7Cl2N3O
256.09
OTBXOEAOVRKTNQ-UHFFFAOYSA-N
6,7-dichloro-2H,3H,5H,10H-imidazo[2,1-b]quinazolin-2-one
ClC1=CC=C2NC3=NC(=O)CN3CC2=C1Cl
68475-42-3
C10H7Cl2N3O
256.09
OTBXOEAOVRKTNQ-UHFFFAOYSA-N
6,7-dichloro-2H,3H,5H,10H-imidazo[2,1-b]quinazolin-2-one
ClC1=CC=C2NC3=NC(=O)CN3CC2=C1Cl

A3978, 68475-42-3

Spectrum™ Anagrelide Hydrochloride, USP
Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 500 g
Water 4.5-7.5%
CAS Max % 1
Percent Purity 98 to 102%