Learn More
Cole-Parmer Texas Electronics 2-200 Analog Wind Speed System, 0 to 100 mph, self-powered  DFS Item
Manufacturer:  Cole-Parmer 9953571
The Model 2-200 Wind Indicating System is an extremely accurate, low cost wind measuring system. This system consists of an anemometer (Model TV-114) and a dial indicator. This system is self-generating; hence no external power source or battery is required. The indicator is contained in a cast aluminum housing with universal mounting brackets. These brackets greatly simplify meter mounting in marine and mobile installations as they provide for top, back, or bottom mounting.
Catalog No. 50-632-953