Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Analytical Precision UltraClass Weight, No Certificate

Combine high precision with the advantage of two-piece construction with the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

Specifications

Includes Sheet Weight, Polycarbonate Case
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Classification UltraClass
Material Stainless Steel
View More Specs

Products 14
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
02-217-026
View Documents
Troemner™
70320
20 mg N/A
02-217-027
View Documents
Troemner™
70310
30 mg N/A
02-217-029
View Documents
Troemner™
70300
50 mg N/A
02-217-030
View Documents
Troemner™
70270
300 mg N/A
02-217-031
View Documents
Troemner™
70230
3 g N/A
02-217-032
View Documents
Troemner™
70200
20 g N/A
02-217-034
View Documents
Troemner™
70160
200 g N/A
02-217-037
View Documents
Troemner™
70150
300 g N/A
02-217-038
View Documents
Troemner™
70140
500 g N/A
02-217-039
View Documents
Troemner™
70130
1 kg N/A
02-217-040
View Documents
Troemner™
70120
2 kg N/A
02-217-043
View Documents
Troemner™
70110
3 kg N/A
02-218-095
View Documents
Troemner™
70090
10 kg N/A
02-218-097
View Documents
Troemner™
70080
20 kg N/A
Description

Description

  • Used in extremely stable environments
  • Two-piece construction with adjusting cavity
  • Supplied in a precision-fit, high quality polycarbonate case
Specifications

Specifications

Sheet Weight, Polycarbonate Case
Calibration of high-precision analytical balances with readability as low as 0.01 mg
Stainless Steel
6.35 cm
7.95 g/cm3 at 20°C
Reference standard calibration
UltraClass
Polycarbonate
5.08 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit