Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ Analytical Precision UltraClass Weight Set, No Certificate
Combine high precision with the advantage of two-piece construction with the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.
Supplier: Troemner™ 30390009
Description
- Used in extremely stable environments
- 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weights 1 g and above are of two-piece construction
- Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
Specifications
1 g and above: Two-piece, 500 mg and below: One-piece | |
UltraClass | |
Polycarbonate | |
8.03 g/cm3 at 20°C | |
8.89 cm | |
Calibration of high-precision analytical balances with readability as low as 0.01 mg | |
11/Set |
No Certificate | |
Troemner Alloy 8 | |
(2) 200 g to 1 g | |
20.32 cm | |
Reference standard calibration | |
Cylindrical Weight: (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, Polycarbonate Case, Forceps, Brush |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction