Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Analytical Precision UltraClass Weight Set, NVLAP Accredited Certificate

Combine high precision with the advantage of two-piece construction with the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

$2050.00 - $5200.00

Specifications

Quantity 13/Set
Includes Cylindrical Weight: 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, Sheet Weight: 500 mg, (2) 200 mg, 100 mg, Polycarbonate Case, Forceps, Brush
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Classification UltraClass
View More Specs

Products 5
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Weight (Metric) Price Quantity & Availability  
02-217-057
View Documents
Troemner™
72250W
100 to 100 mg
Each for $2,340.00
Add to cart
 
02-217-054
View Documents
Troemner™
72220W
200 to 1 g
Each for $2,050.00
Add to cart
 
02-217-051
View Documents
Troemner™
72200W
(2) 200 g to 1 g
Each for $2,270.00
Add to cart
 
02-217-048
View Documents
Troemner™
72190W
500 to 1 mg
Each for $5,200.00
Add to cart
 
02-217-047
View Documents
Troemner™
72180W
500 to 1 g
Each for $3,660.00
Add to cart
 
Description

Description

  • Used in extremely stable environments
  • 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weights 1 g and above are of two-piece construction
  • Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
Specifications

Specifications

13/Set
Reference standard calibration
UltraClass
Polycarbonate
20.32 cm
Cylindrical Weight: 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, Sheet Weight: 500 mg, (2) 200 mg, 100 mg, Polycarbonate Case, Forceps, Brush
Calibration of high-precision analytical balances with readability as low as 0.01 mg
Troemner Alloy 8, Stainless Steel
8.89 cm
Weights 1 g and above are 8.03 g/cm3 at 20°C. Weights below 1 g are 7.95 g/cm3 at 20°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit