Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anandamide (in Tocrisolve 100), Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
94421-68-8
C22H37NO2
347.543
LGEQQWMQCRIYKG-DOFZRALJSA-N
anandamide,arachidonylethanolamide,arachidonoyl ethanolamide,anandamide 20.4, n-6,n-arachidonoyl ethanolamine,arachidonoylethanolamide,n-arachidonoyl-2-hydroxyethylamide,arachidonoyl-ea,aea,n-arachidonoylethanolamine
5281969
CHEBI:2700
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCO
This item is not returnable. View return policy
94421-68-8
C22H37NO2
347.543
LGEQQWMQCRIYKG-DOFZRALJSA-N
anandamide,arachidonylethanolamide,arachidonoyl ethanolamide,anandamide 20.4, n-6,n-arachidonoyl ethanolamine,arachidonoylethanolamide,n-arachidonoyl-2-hydroxyethylamide,arachidonoyl-ea,aea,n-arachidonoylethanolamine
5281969
CHEBI:2700
(5Z,8Z,11Z,14Z)-N-(2-hydroxyethyl)icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCO

Endogenous CB receptor agonist (in water-soluble emulsion)

Endogenous cannabinoid and vanilloid receptor agonist, in water-soluble emulsion (for details see TocrisolveTM 100, Cat. No. 1684). Ki values are 89 and 371 nM for CB1 and CB2 receptors respectively; EC50 values are 18, 31 and 27 nM at GPR55, CB1 and CB2 respectively; pKi = 5.68 for rVR1. Also blocks TNF-α-induced NF-kB activation via direct inhibition of IKK.

Available as the pure oil dissolved in ethanol, Anandamine . Tocrisolve Control also available.

Quantity 10mg
Assay Percent Range >98%
Formula Weight 347.54
Chemical Name or Material Anandamide (in Tocrisolve™ 100)

Recommended Storage : Store at +4°C