Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANAPC11 Mouse anti-Human, Clone: OTI3F5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA506323S
Description
Component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of ′Lys-11′-linked polyubiquitin chains and, to a lower extent, the formation of ′Lys-48′- and ′Lys-63′-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex.Specifications
ANAPC11 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NYG5 | |
ANAPC11 | |
Full length human recombit protein of human ANAPC11 produced in HEk293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3F5 | |
Unconjugated | |
ANAPC11 | |
APC11, Apc11p, HSPC214 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51529 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction