Learn More
Abnova™ ANAPC4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00029945T01
Catalog No. 89-933-361
Add to cart
Description
Description
Specifications
Specifications
293T
NM_013367
pCMV-ANAPC4 full-length
100 μL
Human
ANAPC4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 88.99kDa
Transient overexpression cell lysate was tested with anti-ANAPC4 antibody (Mfr. No. H00029945-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot