Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anastrozole, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1G
5 g
25 g
Packaging:
Amber Glass Bottle
120511-73-1
C17H19N5
293.37
YBBLVLTVTVSKRW-UHFFFAOYSA-N
2-[3-(1-cyano-1-methylethyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]phenyl]-2-methylpropanenitrile
CC(C)(C#N)C1=CC(=CC(CN2C=NC=N2)=C1)C(C)(C)C#N
120511-73-1
C17H19N5
293.37
YBBLVLTVTVSKRW-UHFFFAOYSA-N
2-[3-(1-cyano-1-methylethyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]phenyl]-2-methylpropanenitrile
CC(C)(C#N)C1=CC(=CC(CN2C=NC=N2)=C1)C(C)(C)C#N

A3976, 120511-73-1, C17H19N5

Spectrum™ Anastrozole, USP
Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 25 g
CAS Max % 1
Percent Purity 98 to 102%