Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ancient ubiquitous protein 1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | Ancient ubiquitous protein 1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126197
|
Novus Biologicals
NBP310648100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
Ancient ubiquitous protein 1 Polyclonal specifically detects Ancient ubiquitous protein 1 in Mouse samples. It is validated for Western Blot.Specifications
Ancient ubiquitous protein 1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
ancient ubiquitous protein 1 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse Ancient ubiquitous protein 1 (NP_031543). Peptide sequence MPKDSAFPGAPAALRRWRRQRPRSPEAAAMEPPPAPGPERLFDSHRLPSD | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
550 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title