Learn More
Selleck Chemical LLCSupplier Diversity Partner Andarine 50mg 401900-40-1 GTx-007, S-4  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S114050MG
Andarine (GTx-007, S-4) is a selective non-steroidal androgen receptor (AR) agonist with Ki of 4 nM, tissue-selective for anabolic organs. Phase 3.
Catalog No. 50-688-5