Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andover CoFlex™ LF2 Latex-Free Foam Bandages

Lightweight, nonirritating bandage that provides added patient comfort and prolonged wear

Manufacturer:  Andover Coated Products 9400CP

 View more versions of this product

Catalog No. 22-320-209

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

5 yd.
Self adhesive
Cohesive Bandage
10cm
For all cohesive bandage applications, including the treatment of edema, holding dressings in place, stabilizing IVs, and as a light compression wrap
Foam
Smooth
5 yd. x 4 in. (4.6m x 10cm)
4.6m
Assorted
4 in.
Yes
Blue, light blue, purple, red, neon green, neon pink bandage
Yes
Yes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit