Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Andover CoFlex™ NL Bandages
Latex-free cohesive bandage with EasyTear™ patented technology that can be used to secure, protect, compress, or support
Supplier: Andover 5200BLUE
Description
- EasyTear™ patented technology means no scissors needed
- Soft and strong so it will not slip
- Controlled compression will not constrict
- Easy to apply and remove
- Available in a variety of colors and sizes
Specifications
CoFlex™ NL Bandage | |
5 yd. | |
5 cm | |
5 yd. x 2 in. (4.57 m x 5 cm) | |
Blue | |
Soft and Strong |
Sterile | |
4.57 m | |
2 in. | |
For use on traumatized skin | |
Non-slip | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction