Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andover CoFlex™ NL Kids Pack Bandages

Latex-free cohesive bandage with EasyTear™ patented technology designed with kids in mind

Manufacturer:  Andover Coated Products 5400KP

 View more versions of this product

Catalog No. 19-003-1202


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

5 yd.
Self adhesive
CoFlex™ NL Bandage
10cm
For use on traumatized skin
4.57m
Purple hearts on neon pink, blue dinosaurs on neon green, blue trucks and cars on light blue, bumblebees on yellow
4 in.
Yes
5 yd. x 4 in. (4.57m x 10cm)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit