Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andover CoFlex™ NL Smiley Pack Bandages

Latex-free cohesive bandage with EasyTear™ patented technology designed with a bit of whimsy

Manufacturer:  Andover 5100SC

 View more versions of this product

Catalog No. 22-320-134


Add to cart

Description

Description

  • Smiley faces on Neon Pink, Yellow, Lavender, and Light Blue
  • EasyTear patented technology means no scissors needed
  • Soft and strong so it will not slip
  • Controlled compression will not constrict
  • Easy to apply and remove
  • Available in a variety of sizes

US Patent 5,762,623

Specifications

Specifications

Sterile
4.57m
Smiley on neon pink, yellow, lavender, light blue
1 in.
Yes
Soft and strong
5 yd.
Self adhering and non-slip
CoFlex™ NL Bandage
2.5cm
For use on traumatized skin
5 yd. x 1 in. (4.57m x 2.5cm)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit