Learn More
Androgen Receptor Rabbit anti-Human, KHL, Clone: 3K11, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB115025UL
 View more versions of this product
Catalog No. ZRB115025MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Androgen Receptor Monoclonal antibody specifically detects Androgen Receptor in Human,Rat,Canine,Mouse,Rhesus Macaque samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Androgen Receptor
3K11
KHL
Purified
AR, DHTR, NR3C4
KLH-conjugated Linear Peptide corresponding to the N-terminal of Human Androgen Receptor.
Target Molecular Weight 99.19 kDa calculated; ∼105 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_000035.2
Dihydrotestosterone receptor;Nuclear receptor subfamily 3 group C member 4
Rabbit
IgG
Protein A Purified
Signaling
Primary
Human, Rat, Canine, Mouse, Rhesus Macaque