Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

androgen receptor Rabbit anti-Human, Non-human primate, Porcine, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 225761AP

Catalog No. 50-173-3891


Add to cart

Description

Description

This antibody is a rabbit polyclonal antibody. It can specifically recognize the 110 kDa AR protein.

The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis.AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain.Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy ft.s disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).
Specifications

Specifications

androgen receptor
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10275, Q9GKL7
AIS, androgen receptor, AR, DHTR, Dihydrotestosterone receptor, HUMARA, HYSP1, KD, NR3C4, SBMA, SMAX1, TFM
Rabbit
IgG
150 μL
-20°C
Primary
367, 397582
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
AR
Liquid
AR
androgen receptor Fusion Protein Ag17385
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Non-human primate, Porcine
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit