Learn More
Crystal Chem IncSupplier Diversity Partner Androstenedione ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80952
The Androstenedione ELISA is used to quantify levels of human androstenedione in serum using only a 25 µL sample volume. Highlights: Kit uses sample size of only 25 µL. High sensitivity of 0.05 ng/mL. Dynamic range of 0.1 - 10 ng/mL.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7858