Learn More
BioVision IncSupplier Diversity Partner Angiogenesis (Tube Formation) Assay: Angiogenesis (Tube Formation) Assay: Robust assay to determine angiogenesis (in vitro) and to screen inhibitors & stimulators of angiogenesis in less than 18 hrs. 50 assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K90550
Angiogenesis is the process of generating new blood vessels from the pre-existing vasculature. Angiogenesis is required for growth and development, wound healing, tissue granulation and formation of malignant tumors. The quick assessment of angiogenesis involves measurement of the ability of endothelial cells to form three-dimensional tube-like structures. BioVision’s Tube Formation Assay provides a robust method to determine angiogenesis (in vitro) in less than 18 hrs. This assay kit provides a simple, easy to perform, semi-quantitative tool for assessing angiogenesis. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9972