Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiogenin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF265SP
This item is not returnable.
View return policy
Description
Angiogenin Polyclonal specifically detects Angiogenin in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Angiogenin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALS9, ANG, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
Goat | |
Affinity Purified | |
RUO | |
283 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 1-15 ug/mL | |
Q53X86 | |
ANG | |
E. coli-derived recombinant human Angiogenin Gln25-Pro147 Accession # Q53X86 | |
25 μg | |
Primary | |
Detects human Angiogenin in direct ELISAs and Western blots. | |
Human, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only