Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiogenin Mouse anti-Human, Clone: MM0044-7M15, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB110935120.025MG
Description
Angiogenin Monoclonal specifically detects Angiogenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Angiogenin | |
Monoclonal | |
LYOPH | |
Western Blot 1:100-1000, Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 | |
P03950 | |
ANG | |
human recombinant Angiogenin protein | |
0.025 mg | |
Angiogenesis, Cancer | |
283 | |
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
MM0044-7M15 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This recognizes human Angiogenin in western blot. | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only