Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiogenin Mouse anti-Human, Clone: MM0044-7M15, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$177.16 - $493.00
Specifications
Antigen | Angiogenin |
---|---|
Clone | MM0044-7M15 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11093512
|
Novus Biologicals
NB11093512 |
0.1 mg |
Each for $493.00
|
|
NB018046
|
Novus Biologicals
NB110935120.025MG |
0.025 mg |
Each for $177.16
|
|
Description
Angiogenin Monoclonal specifically detects Angiogenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Angiogenin | |
LYOPH | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer | |
P03950 | |
283 | |
human recombinant Angiogenin protein | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0044-7M15 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
ANG | |
IgG1 | |
Protein G purified | |
This recognizes human Angiogenin in western blot. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title