Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Angiogenin Mouse anti-Human, DyLight 755, Clone: MM0044-7M15, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093512IR
Catalog No. NBT93512IR
Description
Description
Angiogenin Monoclonal antibody specifically detects Angiogenin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Angiogenin
MM0044-7M15
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
ANG
human recombinant Angiogenin protein
Protein G purified
Angiogenesis, Cancer
Primary
283
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This recognizes human Angiogenin in western blot.