Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiogenin Antibody (MM0044-7M15), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11093512F
Description
Angiogenin Monoclonal specifically detects Angiogenin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Angiogenin | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ANG | |
human recombinant Angiogenin protein | |
0.1 mL | |
Primary | |
This recognizes human Angiogenin in western blot. | |
Store at 4C in the dark. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0044-7M15 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer | |
283 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction