Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-1 Mouse anti-Human, Chicken, DyLight 680, Clone: MM0019-26B10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11085464FR
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Angiopoietin-1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ANGPT1 | |
Human recombinant human Angiopoietin-1 | |
0.1 mL | |
Primary | |
No cross reactivity with human Angiopoietin-2 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
MM0019-26B10 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Cell Cycle and Replication | |
284 | |
Human, Chicken | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title