Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-2 Antibody (OTI3H7), Biotin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270173B

Catalog No. NBP270173B


Add to Cart

Description

Description

Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Angiopoietin-2
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
ANGPT2
Recombinant protein fragment corresponding to amino acids 19-496 of human Angiopoietin-2 (NP_001138) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI3H7
Western Blot, Immunohistochemistry
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand
Mouse
Affinity Purified
Angiogenesis, Cancer
285.0
Store at 4°C in the dark.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit