Learn More
Angiopoietin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 246131AP
Catalog No. 50-173-4455
Add to cart
Description
Description
Angiopoietin-2 (ANGPT2) is an antagonist of angiopoietin 1 (ANGPT1) and endothelial TEK tyrosine kinase (TIE-2, TEK). The protein disrupts the vascular remodeling ability of ANGPT1 and may induce endothelial cell apoptosis. Three transcript variants encoding three different isoforms have been found for this gene.
Specifications
Specifications
Angiopoietin 2
0.42 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15123, O35462, O35608
AGPT2, ANG 2, ANG2, angiopoietin 2, ANGPT2
Rabbit
IgG
150 μL
-20°C
Primary
11601, 285, 89805
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ANGPT2
Liquid
ANGPT2
Angiopoietin 2 Fusion Protein Ag17938
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat