Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 1/ANGPTL1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271414AF405

Catalog No. N271414A405


Add to cart

Description

Description

Angiopoietin-like Protein 1/ANGPTL1 Monoclonal antibody specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Angiopoietin-like Protein 1/ANGPTL1
OTI1B2
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ANGPTL1
Recombinant protein fragment corresponding to amino acids 24-491 of human Angiopoietin-like Protein 1/ANGPTL1 (NP_004664) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
ANG-3, ANG3angioarrestin, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, AngY, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162
Mouse
IgG2b
0.1 mL
Primary
9068.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit