Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 1/ANGPTL1, Mouse anti-Human, Clone: 205802, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $304.50

Specifications

Antigen Angiopoietin-like Protein 1/ANGPTL1
Clone 205802
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like Protein 1/ANGPTL1, Gly24-Asp491, Accession # O95841
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10275100
View Documents
R&D Systems
MAB10275100
100 ug
Each for $304.50
Add to cart
 
MAB10275SP
View Documents
R&D Systems
MAB10275SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

Angiopoietin-like Protein 1/ANGPTL1 Monoclonal antibody specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human Samples. It is validated for ELISA.
Specifications

Specifications

Angiopoietin-like Protein 1/ANGPTL1
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like Protein 1/ANGPTL1, Gly24-Asp491, Accession # O95841
Primary
Human
Unconjugated
9068
Purified
ELISA
RUO
205802
Mouse
Monoclonal
IgG2b
ANG-3, ANG3angioarrestin, Angiopoietin like Protein 1, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, ANGPTL1, AngY, AngY1, ARP1, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit